Check Order Status
Porcelain Evaporating Dishes
Actual product may vary
Porcelain Evaporating Dishes
Glazed inside and out except for rim.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FKS13499719
70 mL
FKS13419729
150 mL
FISFB968F
250 mL
FKS13439729
250 mL
FKS13429729
1285 mL
FISFB962M
100 mL
6 Options
Catalog number
FKS13499719
Price (MYR)
681.00
PK
Add to cart
Capacity (Metric):
70 mL
Price (MYR)
681.00
PK
Add to cart
Porcelain dish with lip
Glazed inside and out except for rim
Highly resistant to chemicals
Withstand temperatures up to 1150°C (2102°F)
Without cover
Specifications
Bottom Shape
Round
Capacity (Metric)
70 mL
Material
Porcelain
Shape
Round
Type
Evaporating Dish
For Use With (Application)
Evaporating Processes and Ashing
Interior Finish
Glazed
Temperature Range (English)
2102°F
Temperature Range (Metric)
1150°C
Thickness
2 mm
Height (Metric)
30 mm
Height (English)
1.18 in.
Diameter (English)
2.95 in.
Diameter (Metric)
75 mm