High-Form Porcelain Crucibles
High-Form Porcelain Crucibles
Actual product may vary
High-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F)
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) OuterHeight (Metric)Thickness (Metric)
FKS13499699100 mL65mm54mm2.1mm
FISFB965E30 mL43mm37mm2mm
FKS1344969930 mL43mm37mm2mm
FISFB965G50 mL53mm43mm2.1mm
FKS1348969950 mL53mm43mm2.1mm
FISFB965F40 mL50mm40mm2 mm
FKS1340970940 mL50mm40mm2 mm
Catalog number FKS13499699
Price (MYR)
622.00
PK
Add to cart
Capacity (Metric):
100 mL
Diameter (Metric) Outer:
65mm
Height (Metric):
54mm
Thickness (Metric):
2.1mm
Price (MYR)
622.00
PK
Add to cart
Withstand temperatures up to 1150°C (2102°F)
  • Glazed inside and out except for outside bottom surface and rim
Specifications
MaterialPorcelain
FinishGlazed
Max. Temperature (Metric)1150°C
Thickness (Metric)2.1mm
Weight (Metric)64g
Capacity (Metric)100 mL
Diameter (Metric) Outer65mm
Height (Metric)54mm
Product TypeCrucible
Unit SizePK