Telescopic Electric Stand, ST-P 14/590+500
Telescopic Electric Stand, ST-P 14/590+500
Actual product may vary
Telescopic Electric Stand, ST-P 14/590+500
Use this stand with Polytron™ PT 2500 E, PT 4000 and PT 3100 D and PT 6100 D drives. Kinematica™ Telescopic Electric Stand, ST-P 14/590+500 features an ergonomic design and long lasting, robust construction. Includes electric driven stroke, plate and vessel holder.
Have Questions?
Catalog NumberModelVoltage
FKS13497639ST-P 14/590+500100/120 V
Catalog number FKS13497639
Price (MYR)
37,821.00
EA
Add to cart
Model:
ST-P 14/590+500
Voltage:
100/120 V
Price (MYR)
37,821.00
EA
Add to cart
  • Stand electronics are designed to operate in ambient temperatures of 5 to 40°C and were tested according to EN 60335-1 and UL 962.
  • Ergonomic height adjustment using two buttons
  • Telescopic lift with steel casing
  • Top price-to-performance ratio
  • Also available with vessel holder
  • Powerful integrated lift motor with steel casing
  • Long-lasting, robust construction
  • Excellent stability thanks to solid aluminum base plate
  • Includes non-slip mat
  • Stroke 500mm
Specifications
Product TypeTelescopic Electric Stand
Voltage100/120 V
Length (Metric)500 mm
Bottom TypeStainless steel base plate
For Use With (Application)Designed to operate in ambient temperatures of 5 to 40°C
For Use With (Equipment)Polytron™ PT 2500 E, PT 4000 and PT 3100 D and PT 6100 D drives.
ModelST-P 14/590+500
Unit SizeEA