Polytron™ PT 3100 D Homogenizer Drive Unit
Polytron™ PT 3100 D Homogenizer Drive Unit
Actual product may vary
Polytron™ PT 3100 D Homogenizer Drive Unit
Polytron™ PT 3100 D Homogenizer Drive Unit
Fisherbrand
Polytron™ PT 3100 D Homogenizer Drive Unit
Includes power cord, PC software, RS-232/USB connection cable
Have Questions?
Catalog number FKS13180673
Price (MYR)
65,798.00
EA
Add to cart
Price (MYR)
65,798.00
EA
Add to cart
  • Includes power cord, PC software, RS-232/USB connection cable
Specifications
Speed500 to 30,000 rpm
Voltage230 V
Hertz50/60 Hz
Wattage1200 W
Volume Range (Metric)0.1 mL to 10 L
Dimensions (L x W x H)240 x 130 x 265 mm
Height (Metric)265 mm
Length (Metric)240 mm
Width (Metric)130 mm
DisplayLCD
IncludesUK power cable, PC software, RS-232/USB connection cable
ModelPT 3100 D
Noise Level66 dBA at 25,000 rpm, 73 dBA at 30,000 rpm
Plug TypeUK Plug
Temperature Range (Metric)0°C to 40°C
Weight (Metric)Drive: 3.9 kg
Unit SizeEA