Specific Gravity Hydrometers 200 Series
Specific Gravity Hydrometers 200 Series
Actual product may vary
Specific Gravity Hydrometers 200 Series
Shot loaded specific gravity hydrometers for a variety of surface tension ranges.
Have Questions?
Catalog NumberSpecific Gravity Range
FKS128814281.00 to 1.20 g/mL
Catalog number FKS12881428
Price (MYR)
159.00
EA
Add to cart
Specific Gravity Range:
1.00 to 1.20 g/mL
Price (MYR)
159.00
EA
Add to cart
Compares the relative density of a given liquid with that of water at 15.6°C (60°F). Brannan™ 200 Series Specific Gravity Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
  • 250 mm in length
  • Scale subdivision 0.002
  • Accuracy of ± 0.002 specific gravity
  • Includes protective plastic tube
  • Available adjusted for either low or medium surface tension
  • Available for numerous specific gravity ranges
Specifications
Quantity1Each
TypeSpecific Gravity Hydrometer
Accuracy± 0.002 g/mL
Increments0.002 g/mL
Certifications/ComplianceISO 650 and in the (now superseded) 1960 issue of British Standard 718
Length (Metric)250 mm
Ballast TypeSteel Shot
Baume Range1 to 1.2 g/mL
For Use With (Application)Low surface tension
Specific Gravity Range1.00 to 1.20 g/mL
Temperature of Standardization15.6°C
Unit SizeEA