BS 718 M100 Density Hydrometers
BS 718 M100 Density Hydrometers
Actual product may vary
Fisherbrand
BS 718 M100 Density Hydrometers
For reporting the measured specific gravity of a liquid relative to water. Brannan™ BS 718 M100 Density Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
Have Questions?
Catalog NumberSpecific Gravity RangeIncludes
FKS128791970.800 to 0.900 g/mLPlastic protective tube
Catalog number FKS12879197
Price (MYR)
197.00
EA
Add to cart
Specific Gravity Range:
0.800 to 0.900 g/mL
Includes:
Plastic protective tube
Price (MYR)
197.00
EA
Add to cart
For reporting the measured specific gravity of a liquid relative to water. Brannan™ BS 718 M100 Density Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
  • BS 718 : 1991 is generally aligned with ISO 387 and ISO 649
  • With a range span of 0.100g/mL
  • Available for numerous specific gravity ranges
  • Adjusted at 20°C
  • Includes protective plastic tube
  • Available for both low and medium surface tension
  • 250mm in length
  • Divided every 0.002g/mL
Specifications
Quantity1Pack
TypeM100 Density Hydrometer
Accuracy0.002g/mL
Increments0.002 g/mL
Certifications/ComplianceISO 387, ISO 649
Length (Metric)250 mm
IncludesPlastic protective tube
Ballast TypeSteel Shot
For Use With (Application)Adjusted for low surface tension
Specific Gravity Range0.800 to 0.900 g/mL
Unit SizeEA