Check Order Status
Polyethylene, Aspirator Bottle
Actual product may vary
Fisherbrand
Polyethylene, Aspirator Bottle
Capacity Metric:
25 L
Have Questions?
Catalog number
FKS12785285
Price (MYR)
455.00
EA
Add to cart
Price (MYR)
455.00
EA
Add to cart
Screw on cap
With carry handle
Specifications
Capacity (English)
6.6 gal.
Capacity (Metric)
25 L
Height (Metric) With Closure
460 mm
Material
Polyethylene
Product Type
Aspirator Bottle