Check Order Status
Polyethylene, Aspirator Bottle
Actual product may vary
Fisherbrand
Polyethylene, Aspirator Bottle
Capacity Metric:
10 L
Have Questions?
Catalog number
FKS12765285
Price (MYR)
301.00
EA
Add to cart
Price (MYR)
301.00
EA
Add to cart
Capacity Metric: 10 L
Screw on cap
With carry handle
Specifications
Capacity (English)
2.6 gal.
Capacity (Metric)
10 L
Diameter (Metric) Outer
205 mm
Height (Metric) With Closure
378 mm
Material
Polyethylene
Product Type
Aspirator Bottle