PIPETMANTM TipackTM Filter Tips
PIPETMANTM TipackTM Filter Tips
Actual product may vary
PIPETMANTM TipackTM Filter Tips
PIPETMANTM TipackTM Filter Tips
Fisherbrand
PIPETMANTM TipackTM Filter Tips
Designed to meet the most stringent demands in various fields and applications such as PCR, molecular biology and cell technology. GilsonTM PIPETMANTM TipackTM Filter Tips are designed, developed, and manufactured in order to fit Gilson pipettes perfectly. They play a critical role in preventing aerosol-borne contamination.
Have Questions?
Catalog NumberLength (Metric)Volume (Metric)
FKS126495915.1 cm10 to 100 μL
Catalog number FKS12649591
Price (MYR)
1,156.00
PK
Add to cart
Length (Metric):
5.1 cm
Volume (Metric):
10 to 100 μL
Price (MYR)
1,156.00
PK
Add to cart
  • Optimized geometry and pure polypropylene material ensure complete dispensing for consistent results.
  • Guarantees accuracy and precision.
  • No risk of contamination: Tips are manufactured in a clean room.
  • Gamma ray sterilized: Indicator dots used on Gilson's products undergo an irreversible color change upon sterilization.
  • Guaranteed free of DNA, RNA, DNase, RNase, ATP, Pyrogens and trace metals.
  • Full traceability from the mold to the bench guaranteed.
  • Easy identification with color-coded packaging.
Specifications
Volume (Metric)10 to 100 μL
FilteredFiltered
SterilitySterile
ColorClear
AutoclavableNot Autoclavable
ModelDF100ST
Length (English)5.71 in.
Length (Metric)5.1 cm
Clean ClaimsDNA free, RNA free, RNAse free, DNAse free, ATP free, Endotoxin free
CompatibilityUniversal Fit
FormatRack
Tip StyleStandard
Quantity960 (10x96)
Product TypeAir Displacement
DisposableSingle-use
GraduatedYes
Product LinePipetman Tipack
MaterialPolypropylene
Graduations20 μL; 100 μL
Unit SizePK