Aids the introduction of powder into narrow neck vessels. Pyrex™ Scoop with Hollow Stem sits flat to ensure stability on the balance pan. Available in 3 capacities.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Height (Metric)
FKS12631992
3 mL
70 mm
FKS11970885
6 mL
85 mm
2 Options
Catalog number FKS12631992
Price (MYR)
2,603.00
PK
Add to cart
Capacity (Metric):
3 mL
Height (Metric):
70 mm
Price (MYR)
2,603.00
PK
Add to cart
Aids the introduction of powder into narrow neck vessels. Pyrex™ Scoop with Hollow Stem sits flat to ensure stability on the balance pan. Available in 3 capacities.
With hollow stem to aid introduction of the powder into narrow neck vessels like volumetric flasks
The flat base ensures stability on the balance pan