PYREX™ Heavy-Duty Griffin Beakers
PYREX™ Heavy-Duty Griffin Beakers
Actual product may vary
Fisher Catalogue
PYREX™ Heavy-Duty Griffin Beakers
For use under harsh conditions such as mechanized washing
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS12569995400 mL
FKS119115162000 mL
Catalog number FKS12569995
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
400 mL
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes.
Heavy duty beakers specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. For convenience, the 250 through 4000 mL beakers have a double graduated metric scale to indicate their approximate content. All sizes have an extra large marking spot.
Specifications
Capacity (English)13.52 oz.
Diameter (English) Outer3.22 in.
Diameter (Metric) Outer8.2 cm
Capacity (Metric)400 mL
MaterialBorosilicate Glass
For Use With (Application)Provides The Best Mechanical Strength Under Harsh Conditions Such as Mechanized Washings
Graduation Range25 to 325 mL
Height (English)4.33 in.
Height (Metric)11 cm
Increments25 mL
Label TypeExtra Large Marking
Quantity12 Pk.
TypeBeaker
DisposableReusable
GraduatedYes
Certifications/ComplianceASTM E960
ScaleDual Graduated Metric Scale
StyleGriffin
Unit SizePK