Check Order Status
Tall Form Porcelain Crucibles Without Lid
Actual product may vary
Tall Form Porcelain Crucibles Without Lid
Use these dishes for a variety of applications. Haldenwanger™ Tall Form Porcelain Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FKS12396507
24 mL
FKS12306517
40 mL
2 Options
Catalog number
FKS12396507
Price (MYR)
886.00
PK
Add to cart
Capacity (Metric):
24 mL
Price (MYR)
886.00
PK
Add to cart
Available in a variety of sizes
Sizes up to 175mL glazed except rim
1,000mL and larger are glazed except outside base
Complies with DIN VDE 0335, type C 100
Specifications
Material
Porcelain
Finish
Glazed
Capacity (Metric)
24 mL
Diameter (Metric) Outer
35mm
Height (Metric)
44mm
Product Type
Tall Form Porcelain Crucibles