Borosilicate Glass Butyrometer Tube
Borosilicate Glass Butyrometer Tube
Actual product may vary
Fisher Catalogue
Borosilicate Glass Butyrometer Tube
The Borosilicate Glass Butyrometer Tube is used to determine the fat content of dairy samples using the Gerber method.
Have Questions?
Catalog NumberRange
FKS123789550 to 8%
Catalog number FKS12378955
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Range:
0 to 8%
  • Borosilicate glass
  • Capacity and tolerances in accordance with BS ISO 488:2008 (formerly BS 696)
  • Graduated round stem
Specifications
MaterialBorosilicate Glass
Capacity (Metric)10.94 mL
TypeMilk Butyrometer
For Use With (Application)Determination of fat content using Gerber method
Graduations0.1%
Range0 to 8%
Unit SizeEA