Dispensing Drop Bottles
Dispensing Drop Bottles
Actual product may vary
Dispensing Drop Bottles
Easy-squeeze dispensing bottles allow for critical drop control.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1235821860 mL
Catalog number FKS12358218
Price (MYR)
312.00
PK
Add to cart
Capacity (Metric):
60 mL
Price (MYR)
312.00
PK
Add to cart
  • Sealer cap snaps on easily and continues to hold a tight seal even after repeated use
  • Tip opening is 0.76 mm (0.030 in.)
  • Molded from low density polyethylene (LDPE)
Specifications
Capacity2 oz., 60 mL
Capacity (Metric)60 mL
Capacity (English)2 oz.
ColorTranslucent, Red
MaterialLDPE
No. per Pack12
SterilityNon-sterile
Closure IncludedYes
AutoclavableNot Autoclavable
DisposableReusable
Dropper Capacity2 oz.
Drop Size0.002 oz.
GraduatedNo
MouthNarrow, Threaded
ShapeCylindrical
Height (Metric)109 mm
Product TypeDispensing Bottle
Width (English)1.45 in.
Length (English)4.28 in.
Height (English)4.28 in.
Closure MaterialPolyethylene
Closure Size18 mm
Diameter (Metric)36.8 mm
Closure TypeSnap
Unit SizePK