Round-Base 3.5mL Test Tubes
Round-Base 3.5mL Test Tubes
Actual product may vary
Round-Base 3.5mL Test Tubes
Providing high resistance to centrifugation up to 3000g with a suitable adaptor in a balanced rotor. Gosselin™ Round-Base 3.5mL Test Tubes are available in either polystyrene or polypropylene.
Have Questions?
Catalog NumberMaterialSterilityClosure Type
FKS12357537PolypropyleneNon-sterileNot Included
Catalog number FKS12357537
Price (MYR)
814.00
PK
Add to cart
Material:
Polypropylene
Sterility:
Non-sterile
Closure Type:
Not Included
Price (MYR)
814.00
PK
Add to cart
  • For laboratory use
  • All tubes are CE + IVD marked
  • Available with or without closure
  • Compatible with automation systems
  • Available in sterile or non-sterile (sterility provided by electron beam treatment at 10kGy)
Specifications
Certifications/ComplianceISO 11137
ColorNatural
Capacity (Metric)3.5 mL
MaterialPolypropylene
For Use WithFor clinical, dairy, environment, industries, microbiology
Packaging500BG
Quantity2500 Carton
RimmedNo
SterilityNon-sterile
Diameter (Metric)11.8 mm
Closure TypeNot Included
Product TypeTest Tube
Unit SizePK