Latex Count Drop Tubes
Latex Count Drop Tubes
Actual product may vary
Fisherbrand
Latex Count Drop Tubes
For use as temporary closures when glass blowing
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
FKS1235356940 mmBore size 6 mm
Catalog number FKS12353569
Price (MYR)
499.00
PK
Add to cart
Length (Metric):
40 mm
Diameter (Metric):
Bore size 6 mm
Price (MYR)
499.00
PK
Add to cart
  • Closed at one end for stirring rods
Specifications
For Use With (Application)Temporary closures when glass blowing
Diameter (Metric)Bore size 6 mm
Length (Metric)40 mm
Unit SizePK