Round Bottom Disposable Borosilicate Glass Tubes with Plain End
Round Bottom Disposable Borosilicate Glass Tubes with Plain End
Actual product may vary
Fisherbrand
Round Bottom Disposable Borosilicate Glass Tubes with Plain End
Use these tubes for tissue culture and clinical chemistry applications. Fisherbrand™ Disposable Borosilicate Glass Tubes with Plain End are made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda lime glass.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)Diameter (Metric)
FKS123472794 mL75 mm10 mm
FIS14-961-254 mL75 mm10 mm
FIS14-961-266 mL75 mm12 mm
FIS14-961-2710 mL100 mm13 mm
FIS14-961-2915 mL100 mm16 mm
FIS14-961-3019 mL125 mm16 mm
FIS14-961-3336 mL150 mm20 mm
FKS1150741355 mL150 mm25 mm
Catalog number FKS12347279
Price (MYR)
472.00
PK
Add to cart
Capacity (Metric):
4 mL
Length (Metric):
75 mm
Diameter (Metric):
10 mm
Price (MYR)
472.00
PK
Add to cart
Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda lime glass. These disposable borosilicate glass tubes are perfect for tissue culture and clinical chemistry applications.
  • 10 and 12mm O.D. sizes are available
  • Packaged in shrink-wrapped trays of 125 or 250 to prevent contamination or breakage
Specifications
ColorClear
Design TypePlain
Capacity (Metric)4 mL
Length (Metric)75 mm
MaterialBorosilicate Glass
DisposableSingle-use
For Use WithFor tissue culture and clinical chemistry
PackagingShrink Wrapped Tray
RimmedYes
SterilityNon-sterile
AutoclavableAutoclavable
GraduatedNo
Diameter (Metric)10 mm
Closure TypeKIM-KAP
Product TypeDisposable Culture Tube
Unit SizePK