High impact L-shape disposable bacterial spreaders
Have Questions?
Catalog Number
Quantity
FKS12322048
100 Bags of 5
Catalog number FKS12322048
Price (MYR)
0.00
PK
Add to cart
Quantity:
100 Bags of 5
Price (MYR)
0.00
PK
Add to cart
High impact polystyrene disposable bacterial spreaders. Wedge shaped to reduce tendency to flip, or traditional “L” shape. Packs of 500 contain re-sealable inner bags of either 20 or 5 sterile spreaders.
Specifications
ColorBlue
DisposableSingle-use
Quantity100 Bags of 5
ShapeL-Shape
MaterialPolystyrene
Product TypeSpreader
SterilitySterile
For Use With (Application)High impact Polystyrene disposable bacterial spreaders