Check Order Status
Polypropylene Wadless Screw Caps
Actual product may vary
Fisherbrand
Polypropylene Wadless Screw Caps
Fits wide mouth and Winchester bottles. Azpack™ Polypropylene Wadless Screw Caps are available in a variety of sizes.
Have Questions?
Catalog Number
Color
Neck Finish
FKS12307068
White
R3/24
Catalog number
FKS12307068
Price (MYR)
0.00
PK
Add to cart
Color:
White
Neck Finish:
R3/24
Price (MYR)
0.00
PK
Add to cart
For medical usages
White polypropylene
Softer texture due to white pigmentation
Specifications
Color
White
Material
Polypropylene
Quantity
50/PK
Thread Size (Metric)
TF mm
Neck Finish
R3/24
Product Type
Wadless Screw Cap
For Use With (Equipment)
For BTF-425, 430, 440, 600, 610, 625, 630 series bottles