Azlon™ LDPE Wide Neck Wash Bottles
Azlon™ LDPE Wide Neck Wash Bottles
Actual product may vary
Fisherbrand
Azlon™ LDPE Wide Neck Wash Bottles
Use these translucent plastic bottles for a variety of liquid storage and sampling applications. Azlon™ LDPE Wide Neck Wash Bottles come with color-coded one-piece PP closures that maintain their shape in use.
Have Questions?
Catalog NumberCapacity (Metric)Closure Color
FKS120348271000 mLWhite
Catalog number FKS12034827
Price (MYR)
448.00
PK
Add to cart
Capacity (Metric):
1000 mL
Closure Color:
White
Price (MYR)
448.00
PK
Add to cart
Use these translucent plastic bottles for a variety of liquid storage and sampling applications. AzlonTM LDPE Wide Neck Wash Bottles come with color-coded one-piece PP closures that maintain their shape in use.
  • Square shoulder wash bottles featuring a wide neck for easy spill free filling
  • Fitted with color coded one-piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
Specifications
Capacity (Metric)1000 mL
Capacity (English)33.81 oz.
ColorTranslucent
IncludesWhite Polypropylene closure
MaterialLDPE
Quantity5/PK
Temperature Range (Metric)–50 to 80°C
Closure ColorWhite
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthWide
ShapeRound
Height (Metric)202 mm
Product TypeWash Bottle
Width (English)3.54 in.
Height (English)7.95 in.
Closure MaterialPolypropylene
Diameter (Metric)90 mm
Closure TypeCap
Through the Closure DispensingYes
Unit SizePK