Pyrex™ Gooch Crucible with Integral Glass Sintered Disc
Pyrex™ Gooch Crucible with Integral Glass Sintered Disc
Actual product may vary
Fisherbrand
Pyrex™ Gooch Crucible with Integral Glass Sintered Disc
Ideal for drying precipitates to constant mass at 110°C. Pyrex™ Gooch Crucible includes an integral sintered disc.
Have Questions?
Catalog NumberCapacity (Metric)Porosity
FKS1196102560 mL3
Catalog number FKS11961025
Price (MYR)
1,349.00
PK
Add to cart
Capacity (Metric):
60 mL
Porosity:
3
Price (MYR)
1,349.00
PK
Add to cart
Ideal for drying precipitates to constant mass at 110°C. PyrexTM Gooch Crucible includes an integral sintered disc.
  • Ideal for drying precipitates to constant mass at 110°C
  • Suitable for ignition work up to 450°C
  • With integral glass sintered disc
  • Manufactured from chemically resistant Pyrex™ borosilicate glass
Specifications
DescriptionWith Integral Glass sintered disc
Diameter (Metric) Disc40mm
MaterialBorosilicate Glass
For Use With (Application)Ideal for drying precipitates to constant mass at 110°C and for ignition work up to 450°C
Max. Temperature (Metric)450°C
Capacity (Metric)60 mL
Height (Metric)65mm
Product TypeGooch crucible
Porosity3
Unit SizePK