Borosilicate Glass Graduated Cylinder with Stopper
Borosilicate Glass Graduated Cylinder with Stopper
Actual product may vary
Fisherbrand
Borosilicate Glass Graduated Cylinder with Stopper
Imprinted with blue ceramic graduations for enhanced visibility. Fisherbrand™ Borosilicate Glass Graduated Cylinders with Stopper provide a reliable solution for measuring and mixing liquids in a laboratory.
Have Questions?
Catalog NumberCapacity (Metric)GraduationsHeight (Metric)
FKS11903365100 mL1 mL260 mm
Catalog number FKS11903365
Price (MYR)
362.00
PK
Add to cart
Capacity (Metric):
100 mL
Graduations:
1 mL
Height (Metric):
260 mm
Price (MYR)
362.00
PK
Add to cart
Imprinted with blue ceramic graduations for enhanced visibility. Fisherbrand™ Borosilicate Glass Graduated Cylinders with Stopper provide a reliable solution for measuring and mixing liquids in a laboratory.
  • Features: sturdy hexagonal base, blue ceramic markings, wide range of capacities
  • Hexagonal Stopper: heavy duty blue polypropylene with tight fit for mixing
  • Class B
  • Complies: DIN 12680, BS 604, ISO 4788
Specifications
Base TypeHexagonal
Scale ColorBlue
Certifications/ComplianceISO 4788
ClassClass B
Height (Metric)260 mm
TypeGraduated Measuring Cylinder
MaterialBorosilicate Glass
Diameter (Metric) Outside26 mm
Capacity (Metric)100 mL
IncludesStopper
ScaleBlue Ceramic
Graduations1 mL
AutoclavableAutoclavable
Limit of Error±1 mL
Unit SizePK