Check Order Status
Stainless Steel Forceps
Actual product may vary
Stainless Steel Forceps
For use with filter paper. BOCHEM™ Stainless Steel Forceps have 12mm wide points.
Have Questions?
Catalog Number
Length (Metric)
FKS11835161
125 mm
Catalog number
FKS11835161
Price (MYR)
1,259.00
PK
Add to cart
Length (Metric):
125 mm
Price (MYR)
1,259.00
PK
Add to cart
For use with filter paper
Specifications
No. per Pack
1 Pack
Material
Stainless Steel
Length (Metric)
125 mm
Product Type
Forceps
For Use With (Application)
For filter paper
Width (Metric) Tip
12 mm
Width (Metric)
12 mm
Type
Forceps