Check Order Status
Eutech TN-100 Turbidimeter Calibration Kit
Fisherbrand
Eutech TN-100 Turbidimeter Calibration Kit
Simple, display-prompted push-button calibration
Have Questions?
Catalog number
FKS11786384
Price (MYR)
3,183.00
EA
Add to cart
Price (MYR)
3,183.00
EA
Add to cart
60mL bottles of U.S. EPA-approved, non-formazin standards
Stable and easy to use
Specifications
Type
Calibration Kit Set
Calibration Type
0.02 NTU, 20.0 NTU, 100 NTU, 800 NTU
Includes
Standard solutions; 4 vials of 60mL each
EPA Approved Methods
U.S. EPA-approved non-formazin standards