Grade 55 Ashless Hardened Quantitative Filter Paper
Grade 55 Ashless Hardened Quantitative Filter Paper
Actual product may vary
Fisher Catalogue
Grade 55 Ashless Hardened Quantitative Filter Paper
Suitable for wide range of analytical applications.
Have Questions?
Catalog NumberDiameter (Metric)
FKS11772405125 mm
Catalog number FKS11772405
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
125 mm
  • High wet strength and chemical resistance
  • Made from refined cellulose and cotton linters
  • Washed with acid and cleaned with demineralized water to obtain high purity
  • Low ash, hardened paper with typical ash content of 0.007%
  • Slightly grained surface and suitable for wide range of analytical applications
Specifications
DescriptionAsh Content: 0.007%
Filter TypeHardened Ashless
FormatDisc
MaterialRefined Cellulose, Cotton Linters
Particle Retention2 to 3 μm
Pore Size2 to 3 μm
Filtration Speed180 sec.
For Use With (Application)Used for Acid solutions analysis and for filtration under high pressure
Grade55
Quantity100/Pk.
Thickness (Metric)0.14 mm
TypeQuantitative Filter Paper
Diameter (Metric)125 mm
Unit SizePK