Wheaton™ 47mm Filtration Assembly
Wheaton™ 47mm Filtration Assembly
Actual product may vary
Wheaton™ 47mm Filtration Assembly
Designed to handle up to 1000mL of liquid for analysis of particulate or microbiological contamination
Have Questions?
Catalog NumberIncludes
FKS11758799One stainless steel support with 120 mesh screen, PTFE support screen gasket, stainless-steel support base, 300mL graduated funnel, anodized aluminum clamp, 1L filter flask, No.2 hose connection
Catalog number FKS11758799
Price (MYR)
6,957.00
EA
Add to cart
Includes:
One stainless steel support with 120 mesh screen, PTFE support screen gasket, stainless-steel support base, 300mL graduated funnel, anodized aluminum clamp, 1L filter flask, No.2 hose connection
Price (MYR)
6,957.00
EA
Add to cart
  • For filteration of viscous or proteinaceous solutions to produce ultra clean filtrate
  • Glass components manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
  • Flasks have No.2 hose connection for 0.25 in. (6mm) I.D. tubing
Specifications
ConnectorsNo. 8 stopper
For Use With (Equipment)Filtration Assembly (47 mm)
For Use With (Application)To filter viscous or proteinaceous solutions, to produce an ultra clean filtrate
IncludesOne stainless steel support with 120 mesh screen, PTFE support screen gasket, stainless-steel support base, 300mL graduated funnel, anodized aluminum clamp, 1L filter flask, No.2 hose connection
No. per Case1
TypeFiltration Assembly
Unit SizeEA