Double Open End Bonded Gradient-Density Filter Cartridges
Double Open End Bonded Gradient-Density Filter Cartridges
Actual product may vary
Fisherbrand
Double Open End Bonded Gradient-Density Filter Cartridges
Remove a broad range of particles with these filter cartridges. Cole Palmer™ Double Open End Bonded Gradient-Density Filter Cartridges feature rigid designs ideal for filtering high-solids and higher-viscosity fluids that require higher differential pressures.
Have Questions?
Catalog NumberPore Size
FKS117352991 μm
Catalog number FKS11735299
Price (MYR)
419.00
EA
Add to cart
Pore Size:
1 μm
Price (MYR)
419.00
EA
Add to cart
  • Polypropylene fiber medium is thermally bonded to a rigid core for added durability
  • Cartridges impart no taste, odor or color
  • Suitable to 79°C
  • For optimal performance, change cartridges at 25 psid
Specifications
Flow Rate2 GPM
MaterialPolypropylene
Pore Size1 μm
Filter Size10 in.
For Use With (Application)For removing a broad range of deformable and non-deformable particles, filtering high-solids, higher-viscosity fluids that require higher differential pressures
Temperature (Metric) Max. Operating79°C
TypeLiquid Filter Cartridge
Length (Metric)254 mm
Unit SizeEA