Stainless Steel Chattaway Spatulas
Stainless Steel Chattaway Spatulas
Actual product may vary
Fisher Catalogue
Stainless Steel Chattaway Spatulas
Scrapes and lifts materials from bottles or flasks. Nickel Electro™ Stainless Steel Chattaway Spatulas are non-magnetic with a bright finish. Can be used for sterile applications.
Have Questions?
Catalog NumberLength (Metric)
FKS11726607100 mm
Catalog number FKS11726607
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
100 mm
Scrapes and lifts materials from bottles or flasks. Nickel Electro™ Stainless Steel Chattaway Spatulas are non-magnetic with a bright finish. Can be used for sterile applications.
  • Manufactured from 304 18/8 Stainless Steel
  • Rounded bent end is useful for scraping or lifting materials from bottles or flasks
  • Non-magnetic
  • Bright finish
Specifications
Quantity5 Pk.
SterilitySterile
AutoclavableAutoclavable
Product TypeSpatula
FinishBright
For Use With (Application)For Sterile Applications
Width (Metric) Blade10 mm
Temperature (Metric) Max-196°C to +550°C
MaterialStainless Steel
Length (Metric)100 mm
Unit SizePK