Cell Scrapers
Cell Scrapers
Actual product may vary
Cell Scrapers
Cell Scrapers
Cell Scrapers
Fisherbrand
Cell Scrapers
Easily harvest cells from tissue culture flasks
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English) HandleLength (English) BladeSize
FKS1159769210 in.0.7 in.Small
FIS08-100-24110 in.0.7 in.Small
Catalog number FKS11597692
Price (MYR)
227.00
PK
Add to cart
Length (English) Handle:
10 in.
Length (English) Blade:
0.7 in.
Size:
Small
Price (MYR)
227.00
PK
Add to cart
Easily harvest cells from tissue culture flasks
Fisherbrand Cell Scrapers are appropriate for a variety of cell culture vessels. The uniquely designed handle and blade reduce cell damage during the harvesting process.
  • Presterilized and individually wrapped in peel-open packaging
  • Nonpyrogenic
  • Polyethylene copolymer blade pivots to permit even contact with growth surface
  • Ribbed polystyrene handle provides secure grip and added control
  • Flexible joint between blade and handle eases entry into vessel
  • Pivoting blade improves position control to adjust for difficult angles during cell harvesting
Specifications
Length (English) Blade0.7 in.
Length (Metric) Blade1.8 cm
For Use With (Equipment)For a variety of cell culture vessels
Length (English) Handle10 in.
Length (Metric) Handle25 cm
Blade MaterialPolyethylene Copolymer
Product TypeCell Scraper
SizeSmall
MaterialPolystyrene (Handle)
Unit SizePK