Stainless Steel Chattaway Medium PTFE Coated Spatulas
Stainless Steel Chattaway Medium PTFE Coated Spatulas
Actual product may vary
Stainless Steel Chattaway Medium PTFE Coated Spatulas
Get superior results when you use the stainless steel PTFE-coated spatula for your essential laboratory work. Ideal for Biology, Microbiology and Pharmaceutical Applications, the Fisherbrand™ Stainless Steel Chattaway Spatula Medium PTFE Coated is widely used, sturdy, affordable, and resistant to most solvents.
Have Questions?
Catalog NumberLength (Metric)
FKS11593502200 mm
Catalog number FKS11593502
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
200 mm
  • Resistant to deterioration from contact with boiling water, acids, bases, and most solvents
  • Ideal for use with a wide range of compounds
  • Ideal for Biology, Microbiology and Pharmaceutical Applications
  • Widely used, sturdy, and affordable
Specifications
Quantity1 Ea.
CoatingPTFE Xylan™
Product TypeSpatula
For Use With (Application)Ideal for Biology, Microbiology and Pharmaceutical Applications
Width (Metric) Blade10 mm
Temperature (Metric) Max-195°C to +260°C
MaterialStainless Steel
Length (Metric)200 mm
Unit SizeEA