Color-coded Octahedral PTFE Stir Bar
Color-coded Octahedral PTFE Stir Bar
Actual product may vary
Color-coded Octahedral PTFE Stir Bar
Use when quick visual identification is highly important. Fisherbrand™ Color-coded Octahedral PTFE Stir Bar is available in three colors: blue, red, and yellow.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Color
FKS1156231325 mm8 mmYellow
FKS1151231325 mm8 mmBlue
Catalog number FKS11562313
Price (MYR)
197.00
PK
Add to cart
Length (Metric):
25 mm
Diameter (Metric):
8 mm
Color:
Yellow
Price (MYR)
197.00
PK
Add to cart
  • Color-coded, octagonal stirrer bars
  • For use where identification is of prime importance
  • Note: Colored PTFE coatings are not as inert as pure PTFE
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorYellow
DescriptionPTFE encapsulated Alnico V magnet core
Diameter (English)0.3125 in.
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Length (English)1 in.
Diameter (Metric)8 mm
Length (Metric)25 mm
Quantity5/Pk.
ShapeOctahedral
Unit SizePK