Stainless Steel Chattaway Micro PTFE Coated Spatulas
Stainless Steel Chattaway Micro PTFE Coated Spatulas
Actual product may vary
Fisherbrand
Stainless Steel Chattaway Micro PTFE Coated Spatulas
Get superior results when you use the stainless steel PTFE-coated spatula for your essential laboratory work. Ideal for Biology, Microbiology and Pharmaceutical Applications, the Fisherbrand™ Stainless Steel Chattaway Spatula Micro PTFE Coated is widely used, sturdy, affordable, and resistant to most solvents.
Have Questions?
Catalog NumberLength (Metric)
FKS11533512150 mm
Catalog number FKS11533512
Price (MYR)
122.00
EA
Add to cart
Length (Metric):
150 mm
Price (MYR)
122.00
EA
Add to cart
  • Resistant to deterioration from contact with boiling water, acids, bases, and most solvents
  • Ideal for use with a wide range of compounds
  • Ideal for Biology, Microbiology and Pharmaceutical Applications
  • Widely used, sturdy, and affordable
Specifications
Quantity1 Ea.
CoatingPTFE Xylan™
Product TypeSpatula
For Use With (Application)Ideal for Biology, Microbiology and Pharmaceutical Applications
Width (Metric) Blade4 mm
Temperature (Metric) Max-195°C to +260°C
MaterialStainless Steel
Length (Metric)150 mm
Unit SizeEA