Aggressive Adhesive Multi-Purpose Nylon Labels with Ribbon for M21 Printers
Aggressive Adhesive Multi-Purpose Nylon Labels with Ribbon for M21 Printers
Actual product may vary
Aggressive Adhesive Multi-Purpose Nylon Labels with Ribbon for M21 Printers
Aggressive Adhesive Multi-Purpose Nylon Labels with Ribbon for M21 Printers
Aggressive Adhesive Multi-Purpose Nylon Labels with Ribbon for M21 Printers
Fisherbrand
Aggressive Adhesive Multi-Purpose Nylon Labels with Ribbon for M21 Printers
High adhesion and high pliability are the hallmarks of Brady’s nylon cloth (B-499) label material designed to meet a broad array of identification requirements from control panels to liquid nitrogen environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWidth (English)
FKS115238640.3 in.
FIS19-102-7460.75 in.
Catalog number FKS11523864
Price (MYR)
436.00
BX
Add to cart
Width (English):
0.3 in.
Price (MYR)
436.00
BX
Add to cart
For wire, cable, faceplates and patch panels
  • High adhesion and high pliability are the hallmarks of Brady’s nylon cloth (B-499) label material designed to meet a broad array of identification requirements from control panels to liquid nitrogen environments.
  • Application Versatility — Highly pliable material conforms tightly to curved surfaces such as wires and cables while also adhering to stainless steel, textured ABS, Polypropylene, and many other material types.
  • Engineered Performance — Resists exposure to cold temperatures, chemicals, greases, oils, cleaners, and detergents.
  • Irregular Surfaces — Low profile label material adheres to smooth, rough, or powder-coated surfaces or even materials with low surface energy.
  • Certifications — UL Recognized to UL969, CSA to C22.2 No. 0.15-95, RoHS compliant.
Specifications
AutoclavableAutoclavable
Certifications/ComplianceHalogen-free (DIN VDE 0472/parts 815), UL
DescriptionAggressive Adhesive Multi-purpose Nylon Labels
Adhesive TypePermanent Acrylic
ColorBlack, White
FinishMatte
FormatCartridge (Label)
MaterialNylon Cloth
Print TechnologyThermal Transfer
Product TypeLabel Cartridge
SterilityNon-sterile
For Use With (Application)Communications cables, General identification, Laboratory identification, Panel identification, Voice or data identification, Wire and cable marking
For Use With (Equipment)M210;M210-LAB;M211;BMP21-PLUS;BMP21-LAB;IDPAL;LABPAL
Labels per Row1
Min. Operating Temperature (Metric)-40°C
ModelM21-375-499
No. per Pack1 Tape / Box
Quantity1 Roll
ShapeRectangle
Size9.53 mm (W) x 4.88 m (L)
Size (English) Panel0.375 in W x 16 ft L
Size (Metric) Panel9.53 mm W x 4.88 m L
Temperature Range (English)-40°F to +194°F
Temperature Range (Metric)-40°C to +90°C
Thickness (English)6.5 mil
Thickness (Metric)0.165 mm
Dimensions (LxW)16 ft. x 0.375 in.
Length (English)16 ft.
Length (Metric)4.88 m
Width (English)0.3 in.
Width (Metric)7.62 mm
Height (Metric)4.8 cm
Alternate Part NumberM21-375-499
Unit SizeBX