Check Order Status
Blender Bag Clips
Actual product may vary
Blender Bag Clips
For all homogenizer bags that fit Stomacher machines and other paddle blenders. Fisherbrand™ Blender Bag Clips are manufactured from polyethylene.
Have Questions?
Catalog Number
Material
FKS11522882
Polyethylene
Catalog number
FKS11522882
Price (MYR)
1,882.00
PK
Add to cart
Material:
Polyethylene
Price (MYR)
1,882.00
PK
Add to cart
For all blender type bags
Available in packs of 200
Specifications
Type
Clip
Material
Polyethylene
For Use With (Equipment)
Blender Bag
Color
Natural