Thermax™ 8 Level Irreversible Temperature Strips
Thermax™ 8 Level Irreversible Temperature Strips
Actual product may vary
Thermax™ 8 Level Irreversible Temperature Strips
Have Questions?
Catalog NumberTemperature Range (Metric)
FKS11522513204°C to 260°C
Catalog number FKS11522513
Price (MYR)
-
Temperature Range (Metric):
204°C to 260°C
The standard 8 level range covers 40°C to 260°C for general purpose temperature monitoring.
Specifications
Max. Temperature (Metric)260°C
Min. Temperature (Metric)204°C
Temperature Range (Metric)204°C to 260°C
Unit SizePK