Laboratory Jacks
Laboratory Jacks
Actual product may vary
Laboratory Jacks
Provides for precise height adjustment with the turn of a knob. Fisherbrand™ Laboratory Jacks are manufactured from aluminum alloy with a lifting screw in stainless steel.
Have Questions?
Catalog NumberWidth (Metric) BaseLength (Metric) BaseCapacity (Metric)Height (Metric)
FKS11507023160 mm170 mm25 kg50 to 270 mm (Top Plate)
Catalog number FKS11507023
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Width (Metric) Base:
160 mm
Length (Metric) Base:
170 mm
Capacity (Metric):
25 kg
Height (Metric):
50 to 270 mm (Top Plate)
  • Plastic knob operates the scissor action lifting mechanism
  • Either plate can be used as the base
  • Available in a range of plate dimensions
Specifications
MaterialAluminum
Height (Metric)50 to 270 mm (Top Plate)
Length (Metric) Base170 mm
Width (Metric) Base160 mm
Capacity (Metric)25 kg
IncludesStainless steel Lifting screw
Product TypeLaboratory Lift
For Use With (Equipment)12mm dia. Support Rod
Quantity1 Ea.
Unit SizeEA