Grade 11 Ashless Fast Filtering Quantitative Filter Paper
Grade 11 Ashless Fast Filtering Quantitative Filter Paper
Actual product may vary
Fisher Catalogue
Grade 11 Ashless Fast Filtering Quantitative Filter Paper
Used for coarse and voluminous precipitates such as iron, aluminum and chromium hydroxides, Si-determination in steel and pig iron analysis.
Have Questions?
Catalog NumberFormatDiameter (Metric)
FKS11475188Disc110 mm
Catalog number FKS11475188
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Format:
Disc
Diameter (Metric):
110 mm
  • Wide pores filter paper
  • Made with 100% cotton linter fibers
  • Washed with acid and cleaned with demineralized water to obtain high purity
  • Ash content is 0.007%
  • Applications: Chemical, food, beverage industries and atmospheric analysis
Specifications
DescriptionAsh Content: 0.007%
Filter TypeAshless
FormatDisc
MaterialCotton Linters
Particle Retention12 to 15 μm
Pore Size12 to 15 μm
Filtration Speed10 sec.
For Use With (Application)Used for coarse and voluminous precipitates such as Iron, Aluminum and Chromium Hydroxide, Si determination in Steel and Pig Iron analysis
Grade11
Quantity100/Pk.
Thickness (Metric)0.18 mm
TypeQuantitative Filter Paper
Diameter (Metric)110 mm
Unit SizePK