SHP-200-MP Stuart Three Position Stirring Hotplate
SHP-200-MP Stuart Three Position Stirring Hotplate
Actual product may vary
Fisherbrand
SHP-200-MP Stuart Three Position Stirring Hotplate
SHP-200-MP Stuart Three Position Stirring Hotplate has three independent heating/stirring positions yet only measures 23-5/8 in. L x 10-5/8 in. W (60 x 27 cm).
Have Questions?
Catalog NumberTop Plate Material
FKS11436538Aluminum
Catalog number FKS11436538
Price (MYR)
18,713.00
EA
Add to cart
Top Plate Material:
Aluminum
Price (MYR)
18,713.00
EA
Add to cart
  • Powerful magnets for strong magnetic coupling and stirring capacity
  • Easily accommodate a 2-L beaker in each position
  • Aluminum/silicon alloy top plate offers even plate temperature
  • A flashing Hot warning light indicates temperature above 50°C
Three independently controlled position units only require one power source. Run three separate tests at the same time or run multiple samples under the same conditions.
Specifications
Top Plate MaterialAluminum
Voltage230 V
Includes(3) 25 mm PTFE-Coated Stir Bars
Safety FeaturesSeparate Hot warning lights for each plate
Stirring Features3 Independent heating/stirring positions in a footprint only 600 x 270 mm, Easily accommodates 3 x 2 liter beakers
Electrical Requirements230 V, 50 Hz, 2250 W
Controller TypeAnalog
ModelSB162-3
Sole FeaturesUnit only requires one power point and is ideal for quality control applications where multiple samples require simultaneous heating and stirring, under the same conditions
Size160 x 160 mm
Height (Metric)110 mm
Temperature (Metric)Ambient to 325°C
Height (English)4.3 in.
Frequency50 Hz
Width (Metric)600 mm
Width (English)23.6 in.
Wattage2250 W
Weight (English)24.3 lb.
Weight (Metric)11 kg
TypeStirring Hotplate
ShapeSquare
No. of Positions3
Max. Container Size15 L
Stirring Range100 to 1500 rpm
Unit SizeEA