Search Thermo Fisher Scientific
| Catalog Number | Description | Absorbancy | Quantity |
|---|---|---|---|
| FKS11399183 | BenchGuard Extra | 800 mL/m2 | 50 x Sheets |
| BG50 | BenchGuard | 400 mL/m2 | 1 x 50m Roll |
| FKS11359183 | BenchGuard | 400 mL/m2 | 1 x 50m Roll |
| BG50E | BenchGuard Extra | 800 mL/m2 | 1 x 50m Roll |
Protect benches and other surfaces against liquid spills with the highly absorbent Thermo Scientific™ Sterilin™ BenchGuard. Absorbent liner also provides a cushion for glassware when accidentally dropped on benchtop.