Pyrex™ Borosilicate Glass Class A Volumetric Flask
Pyrex™ Borosilicate Glass Class A Volumetric Flask
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Class A Volumetric Flask
Manufactured from chemically resistant Pyrex™ Borosilicate Glass. Pyrex™ Borosilicate Glass Class A Volumetric Flask includes white enamel inscriptions for easy volume assessment and a polyethylene stopper. Complies with ISO 1042 and DIN 12664, is calibrated to the tolerance required by the United States Pharmacopoeia (USP,) and Conformity 'H' certified.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeHeight (Metric)Tolerance
FKS11393624500 mL19/26260 mm±0.2 mL
FKS11383624250 mL14/23220 mm±0.12 mL
FKS1135362450 mL12/21140 mm±0.05 mL
Catalog number FKS11393624
Price (MYR)
646.00
PK
Add to cart
Capacity (Metric):
500 mL
Closure Size:
19/26
Height (Metric):
260 mm
Tolerance:
±0.2 mL
Price (MYR)
646.00
PK
Add to cart
Manufactured from chemically resistant Pyrex™ Borosilicate Glass. Pyrex™ Borosilicate Glass Class A Volumetric Flask includes white enamel inscriptions for easy volume assessment and a polyethylene stopper. Complies with ISO 1042 and DIN 12664, is calibrated to the tolerance required by the United States Pharmacopoeia (USP,) and Conformity 'H' certified.
  • Complies with ISO 1042 and DIN 12664
  • Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which is tighter than those of ISO/DIN standards
  • Each flask is laser etched with an individual serial number batch code and date of manufacture
  • Batch certificate available to download online
  • Conformity 'H' certified
  • Manufactured from chemically resistant Pyrex™ Borosilicate Glass
  • Inscriptions in white enamel
  • Polyethylene stopper
Specifications
Certifications/ComplianceISO 1042, DIN 12664, USP, Conformity 'H' certified
Closure MaterialPolyethylene
DescriptionWhite enamel inscriptions
Capacity (Metric)500 mL
MaterialBorosilicate Glass
Height (Metric)260 mm
Tolerance±0.2 mL
GraduatedYes
ClassA
Closure Size19/26
Unit SizePK