Premium Microcentrifuge Tubes: 2.0mL
Premium Microcentrifuge Tubes: 2.0mL
Actual product may vary
Premium Microcentrifuge Tubes: 2.0mL
Premium Microcentrifuge Tubes: 2.0mL
Fisherbrand
Premium Microcentrifuge Tubes: 2.0mL
Flat-top snap caps provide a safe, liquid-tight, reliable seal even with prolonged boiling
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS11393613Natural
FIS05-408-138Natural
Catalog number FKS11393613
Price (MYR)
113.00
PK
Add to cart
Color:
Natural
Price (MYR)
113.00
PK
Add to cart
Flat-top snap caps provide a safe, liquid-tight, reliable seal even with prolonged boiling
Capacity 2.0mL
  • High-clarity polypropylene
  • Graduated in 0.25mL increments
  • Withstand forces up to 30,000 × g
  • Easy-open caps with needle insertion spot and textured marking area
  • Labeling area on tube exterior
  • Highly polished interior for maximum sample recovery
  • Temperature range of -80° to +121°C
  • Fit all standard rotors
  • Nonpyrogenic
  • Autoclavable at 121°C/15psi for 15 minutes
Specifications
Cap StyleSnap Cap with Flat Top
Certifications/ComplianceRNase, DNase-free
AutoclavableAutoclavable
Capacity (Metric)2 mL
ColorNatural
Length (Metric)40.6 mm
MaterialPolypropylene
IncludesNeedle insertion spot
Label TypeLabeling Area
GraduatedYes
Closure TypeFlat Snap Cap
Max. RCF30,000 x g
Diameter (Metric)11 mm
Unit SizePK