Whatman™ Clear Polystyrene Universal Lid
Whatman™ Clear Polystyrene Universal Lid
Actual product may vary
Fisherbrand
Whatman™ Clear Polystyrene Universal Lid
Lids are suitable for using as dust covers and to prevent splashing or contamination when plates are being moved around the laboratory.
Have Questions?
Catalog number FKS11388694
Price (MYR)
2,182.00
PK
Add to cart
Price (MYR)
2,182.00
PK
Add to cart
Specifications
ColorClear
MaterialPolystyrene
No. per Pack100 Pieces
For Use With (Application)Lids are suitable for using as dust covers and to prevent splashing or contamination when plates are being moved around the laboratory.
Product TypeUniversal Lid
Unit SizePK