Rota Rack™ Large Tube Rack
Rota Rack™ Large Tube Rack
Actual product may vary
Fisherbrand
Rota Rack™ Large Tube Rack
Use several sizes of large test tubes at once. Rota Rack™ Large Tube Rack has four modules that can be independently rotated to accommodate different sizes. Just click each module into place and load it with tubes.
Have Questions?
Catalog number FKS11384085
Price (MYR)
573.00
EA
Add to cart
Price (MYR)
573.00
EA
Add to cart
Just twist and lock.
  • Each module can be independently rotated.
  • Use different sizes of test tubes at the same time.
  • Click each module into place and load with tubes.
  • Rack can be securely carried and are fully autoclavable.
  • Racks remain fully orientated until they are twisted again.
Four modules each hold four sizes.
  • 2 x 50 mL tubes
  • 8 x 15 mL tubes
  • 6 x 16 mm to 20 mm tubes
  • 10 x 10 mm to 13 mm tubes
Each sturdy polypropylene rack has modules in green, purple, blue and yellow.
Specifications
Product TypeUniversal Multi Rack
AutoclavableAutoclavable
ColorAssorted
HoldsTubes (10 x 10 to 13 mL, 6 x 15 to 20 mL, 8 x 10 to 15 mL, 2 x 50 mL)
MaterialPolypropylene
Positions2 x 50 mL Tubes, 6 x 16 mL to 20 mL Tubes, 8 x 15 mL Tubes, or 10 x 10 mL to 13 mL Tubes
ShapeRectangular, Rounded Corners
ShelvesNone
StackableYes, When Empty
SterilityNon-sterile
For Use With (Equipment)For Use With Large Tubes
Quantity1 Ea.
SizeLarge
Length (Metric)224 mm
Length (English)8.8 in.
Width (Metric)110 mm
Width (English)4.3 in.
Height (Metric)110 mm
Height (English)4.3 in.
Weight (English)2.2 lb.
Weight (Metric)1 kg
Unit SizeEA