Pyrex™ Borosilicate Glass Baffled Erlenmeyer Flasks
Pyrex™ Borosilicate Glass Baffled Erlenmeyer Flasks
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Baffled Erlenmeyer Flasks
Contains four indents to mix contents consistently and without risk of spillage. Pyrex™ Borosilicate Glass Baffled Erlenmeyer Flasks can withstand multiple autoclave cycles.
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric) Outer
FKS113834542000 mL282 mm161 mm
Catalog number FKS11383454
Price (MYR)
1,130.85
Each
Add to cart
Capacity (Metric):
2000 mL
Height (Metric):
282 mm
Diameter (Metric) Outer:
161 mm
Price (MYR)
1,130.85
Each
Add to cart
Contains four indents to mix contents consistently and without risk of spillage. Pyrex™ Borosilicate Glass Baffled Erlenmeyer Flasks can withstand multiple autoclave cycles.
  • Ideal for use in preparation of cultures
  • Manufactured from Pyrex™ Borosilicate Glass
  • Exhibits 4 baffle indents to provide optimal and consistent agitation
  • Suitable for repeated autoclave cycles at 121°C
Specifications
DescriptionConical; Style: Erlenmeyer
Capacity (Metric)2000 mL
MaterialBorosilicate Glass, Borosilicate Glass
For Use With (Application)Preparation of cultures
Height (Metric)282 mm
Neck StyleNarrow
AutoclavableAutoclavable
Mouth StyleNarrow
Diameter (Metric) Outer161 mm
Unit SizeEach