96-Well DeepWell™ Polypropylene Microplates
96-Well DeepWell™ Polypropylene Microplates
Actual product may vary
96-Well DeepWell™ Polypropylene Microplates
96-Well DeepWell™ Polypropylene Microplates
96-Well DeepWell™ Polypropylene Microplates
96-Well DeepWell™ Polypropylene Microplates
Fisherbrand
96-Well DeepWell™ Polypropylene Microplates
Ideal for storage because of the low-binding characteristics of polypropylene.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityVolume (Metric) WellVolume (Metric) Working
FKS11381555Non-sterile1300 μL1000 μL
FIS12-566-120Non-sterile1300 μL1000 μL
Catalog number FKS11381555
Price (MYR)
1,896.00
PK
Add to cart
Sterility:
Non-sterile
Volume (Metric) Well:
1300 μL
Volume (Metric) Working:
1000 μL
Price (MYR)
1,896.00
PK
Add to cart
Ideal for storage because of the low-binding characteristics of polypropylene
  • Accommodate even larger working volumes; up to 1.0 mL or 2.0 mL storage volume per well
  • Proteins and DNA will not adhere to PP, allowing for complete sample recovery.
  • Natural color
  • Nonsterile and sterile available
Specifications
FormatStandard
LidWithout Lid
Array8 x 12
Packaging50 Cases
ColorNatural
Product TypeMicroplate
MaterialPolypropylene
Volume (Metric) Well1300 μL
No. of Wells96
Volume (Metric) Working1000 μL
SterilityNon-sterile
Well ShapeRound
For Use With (Application)For storage of low-binding characteristics of polypropylene
Quantity50/Cs.
Unit SizePK