Whatman™ Benchkote™ Surface Protector Reel
Whatman™ Benchkote™ Surface Protector Reel
Actual product may vary
Whatman™ Benchkote™ Surface Protector Reel
Minimize mess and contamination during spills. Whatman™ Benchkote material has a high-quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.
Have Questions?
Catalog NumberDimensions (L x W)
FKS11380664460mm x 50m
Catalog number FKS11380664
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Dimensions (L x W):
460mm x 50m
Minimize mess and contamination during spills. Whatman™ Benchkote material has a high-quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.
  • Strong: Benchkote surface protector is strong and tear resistant when wet or dry
  • Ease-of-use: Smooth white surface can be written on with ink or pencil and lies flat
  • Clean: Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
  • Disposable: Benchkote paper can be incinerated after use
  • Safe combustion: Polyethylene layer is rapidly consumed when incinerated and does not melt or drip
Specifications
Product TypeSurface Protector
MaterialPolyethylene
Absorbancy750 mL/m2
Length (English)18 in.
Length (Metric)460 mm
Width (English)164 ft.
Width (Metric)50 m
Dimensions (L x W)460mm x 50m
For Use With (Application)Suitable for saturation with disinfectant
Quantity1 Piece
Unit SizeEA