Azlon™ Polypropylene Graduated Molded Jugs with Handles
Azlon™ Polypropylene Graduated Molded Jugs with Handles
Actual product may vary
Fisherbrand
Azlon™ Polypropylene Graduated Molded Jugs with Handles
Manufactured in high clarity polypropylene. Azlon™ Polypropylene Graduated Moulded Jugs with Handles are tapered with ergonomically designed handle for comfort and control. Suitable for food applications in accordance with EU and US regulations. Permanent moulded graduations provide easy volume assessment. Autoclavable to 121° C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS113736945000 mL500mL
FKS11333694500 mL100mL
FKS113436941000 mL100mL
FKS113536942000 mL500mL
Catalog number FKS11373694
Price (MYR)
151.00
EA
Add to cart
Capacity (Metric):
5000 mL
Graduations:
500mL
Price (MYR)
151.00
EA
Add to cart
Manufactured in high clarity polypropylene. Azlon™ Polypropylene Graduated Moulded Jugs with Handles are tapered with ergonomically designed handle for comfort and control. Suitable for food applications in accordance with EU and US regulations. Permanent moulded graduations provide easy volume assessment. Autoclavable to 121° C.
  • Tapered jugs in high clarity polypropylene
  • Permanent molded graduations
  • Autoclavable at 121° C
  • Ergonomically designed handle with thumb grip for easy carrying
Specifications
Capacity (Metric)5000 mL
Capacity (English)169.07 oz.
ColorClear
MaterialPolypropylene
No. per PackIndividual
QuantityEach
Temperature Range (Metric)–20 to 135°C
Graduations500mL
Certifications/ComplianceEU; USA
Closure IncludedNo
AutoclavableAutoclavable
GraduatedYes
MouthWide
ShapeTapered
Height (Metric)270 mm
Product TypeMolded Jug
Height (English)10.63 in.
Diameter (Metric)200 mm
Through the Closure DispensingNo
Unit SizeEA