Polypropylene Tall Form Measuring Jugs
Polypropylene Tall Form Measuring Jugs
Actual product may vary
Polypropylene Tall Form Measuring Jugs
Manufactured from polypropylene for high thermal resistance. Kartell™ Measuring Jugs are transparent with permanent molded graduations for accurate measurements.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS113585031000 mL20/1mL
Catalog number FKS11358503
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Graduations:
20/1mL
  • Convenient integral spout and handle allow easy dispensing
  • Autoclavable
  • Suitable for foodstuff
  • Tall form saves space in the laboratory
Specifications
Capacity (Metric)1000 mL
ColorTransparent
For Use With (Application)Foodstuff
FormTall form
MaterialPolypropylene
Graduations20/1mL
AutoclavableAutoclavable
GraduatedYes
Height (Metric)169 mm
Product TypeMeasuring Jug
Diameter (Metric)117 mm
Unit SizeEA