Pyrex™ Labware Class AS Borosilicate Glass Burette
Pyrex™ Labware Class AS Borosilicate Glass Burette
Actual product may vary
Fisherbrand
Pyrex™ Labware Class AS Borosilicate Glass Burette
Manufactured from precision-bore tubing (borosilicate glass) for high accuracy and complies with ISO 385 standards. Pyrex™ Labware Class AS Borosilicate Glass Buret includes grease-free interchangeable PTFE key with 1.5mm bore. Graduations in whit
Have Questions?
Catalog NumberCapacity (Metric)
FKS1131796450 mL
Catalog number FKS11317964
Price (MYR)
512.00
EA
Add to cart
Capacity (Metric):
50 mL
Price (MYR)
512.00
EA
Add to cart
Manufactured from precision-bore tubing (borosilicate glass) for high accuracy and complies with ISO 385 standards. Pyrex™ Labware Class AS Borosilicate Glass Buret includes grease-free interchangeable PTFE key with 1.5mm bore. Graduations in whit
  • Complies with ISO 385
  • Manufactured from precision-bore tubing (borosilicate glass) for high accuracy
  • Each burette has a +30s waiting time before final reading can be taken
  • Graduations in white enamel for increased visibility with dark solutions
  • Grease free interchangeable PTFE key with 1.5mm bore
Specifications
Capacity (Metric)50 mL
Length (Metric)820 mm
MaterialBorosilicate Glass
Graduations0.1 mL
IncludesInterchangeable PTFE Stopcock Key with 1.5 mm Bore
ScaleWhite
Stopcock MaterialPTFE
Tolerance±0.05 mL
TypeBuret
ClassClass AS
Certifications/ComplianceISO 385
Unit SizeEA