Borosilicate Glass Erlenmeyer Flasks with Screw Cap
Borosilicate Glass Erlenmeyer Flasks with Screw Cap
Actual product may vary
Fisher Catalogue
Borosilicate Glass Erlenmeyer Flasks with Screw Cap
Store media and cultures securely with a screw cap closure. Pryex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap resist thermal shock and chemical corrosion.
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric) OuterClosure Size
FKS107930321000 mL210 mm131 mm42
Catalog number FKS10793032
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Height (Metric):
210 mm
Diameter (Metric) Outer:
131 mm
Closure Size:
42
Store media and cultures securely with a screw cap closure. Pryex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap resist thermal shock and chemical corrosion.
  • PBT screw caps with PTFE faced sealing disc
  • Erlenmeyer pattern
  • Manufactured from Pyrex™ Borosilicate Glass
  • Highest resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
  • Graduated to full capacity
Specifications
Closure MaterialSVL™ Phenolic, PTFE faced with Rubber disc
Capacity (Metric)1000 mL
MaterialBorosilicate Glass, Borosilicate Glass
Height (Metric)210 mm
ShapeConical
GraduatedYes
Closure TypeScrew Cap
Diameter (Metric) Outer131 mm
Closure Size42
Unit SizeEA